2-Benzyloxy-6-fluorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Benzyloxy)-6-fluorophenyl)boronic acid

PC412011-1G
APOSPC412011-1GEA 59.6 EUR
APOSPC412011-1G
2-Benzyloxy-6-fluorphenylboronsäure
2-Benzyloxy-6-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246,0420 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617256
CAS-Nummer: 1217500-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR