(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid 98%

Lieferant: Apollo Scientific

PC412029-5G
APOSPC412029-5GEA 63.8 EUR
APOSPC412029-5G
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid 98%
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid
Formel: C₈H₉BClFO₃
Molecular Weight: 218,4177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15144702
CAS-Nummer: 909122-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR