2-Fluoro-3-methoxyphenylboronic acid, pinacol ester 95+%

Lieferant: Apollo Scientific

PC412056-1G
APOSPC412056-1GEA 21.5 EUR
APOSPC412056-1G
2-Fluoro-3-methoxyphenylboronic acid, pinacol ester 95+%
2-Fluoro-3-methoxyphenylboronic acid, pinacol ester
Formel: C₁₃H₁₈BFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16618962
CAS-Nummer: 1165936-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR