4-Benzyloxy-3,5-difluorophenylboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (4-(Benzyloxy)-3,5-difluorophenyl)boronic acid

PC412068-1G
APOSPC412068-1GEA 68.3 EUR
APOSPC412068-1G
4-Benzyloxy-3,5-difluorophenylboronic acid 97%
4-Benzyloxy-3,5-difluorophenylboronic acid
Formel: C₁₃H₁₁BF₂O₃
Molecular Weight: 264,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16295234
CAS-Nummer: 156635-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR