4-(Trifluormethyl)phenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific

PC412070-5G
APOSPC412070-5GEA 0 EUR
APOSPC412070-5G
4-(Trifluormethyl)phenylboronsäurepinakolester 97%
4-(Trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05863924
CAS-Nummer: 214360-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR