4-(Dimethylcarbamoyl)-2-fluorphenylboronsäure 95%

Lieferant: Apollo Scientific

Synonyme: 4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid

PC412105-1G
APOSPC412105-1GEA 495 EUR
APOSPC412105-1G
4-(Dimethylcarbamoyl)-2-fluorphenylboronsäure 95%
4-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11855833
CAS-Nummer: 874289-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR