3-Benzyloxy-2-chlor-6-fluorphenylboronsäure 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Benzyoxy-2-chloro-6-fluorophenylboronic acid

PC412115-5G
APOSPC412115-5GEA 144 EUR
APOSPC412115-5G
3-Benzyloxy-2-chlor-6-fluorphenylboronsäure 95+%
3-Benzyloxy-2-chlor-6-fluorphenylboronsäure
Formel: C₁₃H₁₁BClFO₃
Molecular Weight: 280,4900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878328
CAS-Nummer: 957062-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR