7-Fluor-2-methylchinolin-8-boronsäure

Lieferant: Apollo Scientific

PC412121-1G
APOSPC412121-1GEA 250 EUR
APOSPC412121-1G
7-Fluor-2-methylchinolin-8-boronsäure
7-Fluor-2-methylchinolin-8-boronsäure
Formel: C₁₀H₉BFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11504838
CAS-Nummer: 1072945-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR