5-Ethyl-2-fluorophenylboronic acid

Lieferant: Apollo Scientific
Achtung

PC412139-1G
APOSPC412139-1GEA 277 EUR
APOSPC412139-1G
5-Ethyl-2-fluorophenylboronic acid
5-Ethyl-2-fluorophenylboronic acid
Formel: C₈H₁₀BFO₂
Molecular Weight: 167,973 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18783131
CAS-Nummer: 900175-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR