5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-(Trifluoromethyl)-1H-pyrazol-4-yl)boronic acid
PC412168-1G
APOSPC412168-1GEA
431
EUR
APOSPC412168-1G
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid 97%
5-Trifluoromethyl-1H-pyrazol-4-ylboronic acid
Formel:
C₄H₄BF₃N₂O₂ Molecular Weight: 179,893 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD07375369 CAS-Nummer: 1202054-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...