2-Fluoro-5-isopropylphenylboronic acid

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-5-isopropylphenyl)boronic acid

PC412198-1G PC412198-250MG
APOSPC412198-1GEA 430 EUR
APOSPC412198-1G APOSPC412198-250MG
2-Fluoro-5-isopropylphenylboronic acid
2-Fluoro-5-isopropylphenylboronic acid
Formel: C₉H₁₂BFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17015760
CAS-Nummer: 1256354-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR