4-Amino-3-fluorophenylboronic acid hydrochloride

Lieferant: Apollo Scientific

Synonyme: (4-Amino-3-fluorophenyl)boronic acid hydrochloride (1:1), 4-Amino-3-fluorophenylboronic acid, HCl, Boronic acid, B-(4-amino-3-fluorophenyl)-, hydrochloride (1:1)

PC412221-1G
APOSPC412221-1GEA 188 EUR
APOSPC412221-1G
4-Amino-3-fluorophenylboronic acid hydrochloride
4-Amino-3-fluorophenylboronic acid hydrochloride
Formel: C₆H₇BFNO₂·HCl
Molecular Weight: 191,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11867880
CAS-Nummer: 1256355-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR