4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

PC412272-1G PC412272-5G PC412272-10G
APOSPC412272-1GEA 120 EUR
APOSPC412272-1G APOSPC412272-5G APOSPC412272-10G
4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid 98%
4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid
Formel: C₁₂H₁₉BF₂O₃Si
Lagertemperatur: Kühlschrank
MDL: MFCD18914502
CAS-Nummer: 870646-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR