3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

PC412317-1G PC412317-250MG
APOSPC412317-1GEA 295 EUR
APOSPC412317-1G APOSPC412317-250MG
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester
Formel: C₁₁H₁₄BClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16995268
CAS-Nummer: 1220219-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR