3,5-Difluor-4-hydroxyphenylboronsäure
Lieferant: Apollo Scientific
Synonyme:
(3,5-Difluoro-4-hydroxyphenyl)boronic acid
PC412357-1G
APOSPC412357-1GEA
295
EUR
APOSPC412357-1G
3,5-Difluor-4-hydroxyphenylboronsäure
3,5-Difluor-4-hydroxyphenylboronsäure
Formel:
C₆H₅BF₂O₃ Molecular Weight: 173.91 g/mol |
MDL:
MFCD17011981 CAS-Nummer: 1132666-81-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...