2,3-Difluoro-4-(methoxycarbonyl)phenylboronic acid
Lieferant: Apollo Scientific
Synonyme:
(2,3-Difluoro-4-(methoxycarbonyl)phenyl)boronic acid
PC412360-250MG
APOSPC412360-250MGEA
340
EUR
APOSPC412360-250MG
2,3-Difluoro-4-(methoxycarbonyl)phenylboronic acid
2,3-Difluoro-4-(methoxycarbonyl)phenylboronic acid
Formel:
C₈H₇BF₂O₄ Molecular Weight: 215,947 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18434428 CAS-Nummer: 1402238-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...