4-Difluoromethoxyphenylboronic acid, pinacol ester 95+%

Lieferant: Apollo Scientific

PC412369-250MG
APOSPC412369-250MGEA 42.2 EUR
APOSPC412369-250MG
4-Difluoromethoxyphenylboronic acid, pinacol ester 95+%
4-Difluoromethoxyphenylboronic acid, pinacol ester
Formel: C₁₂H₁₅BF₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09878543
CAS-Nummer: 887757-48-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR