1-(Difluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Lieferant: Apollo Scientific
Achtung

PC412371-5G PC412371-25G
APOSPC412371-5GEA 59.6 EUR
APOSPC412371-5G APOSPC412371-25G
1-(Difluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-(Difluoromethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel: C₁₀H₁₅BF₂N₂O₂
Molecular Weight: 244.05 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17011852
CAS-Nummer: 1206640-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR