2,3-Difluoropyridine-4-boronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,3-Difluoropyridin-4-yl)boronic acid

PC412378-5G
APOSPC412378-5GEA 154 EUR
APOSPC412378-5G
2,3-Difluoropyridine-4-boronic acid 97%
2,3-Difluoropyridine-4-boronic acid
Formel: C₅H₄BF₂NO₂
Molecular Weight: 158.899 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12022339
CAS-Nummer: 1263374-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR