4-(Dihydroxyboranyl)-2,3-difluorobenzoic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Borono-2,3-difluorobenzoic acid, 4-Carboxy-2,3-difluorophenylboronic acid

PC412389-1G
APOSPC412389-1GEA 111 EUR
APOSPC412389-1G
4-(Dihydroxyboranyl)-2,3-difluorobenzoic acid 95+%
4-(Dihydroxyboranyl)-2,3-difluorobenzoic acid
Formel: C₇H₅BF₂O₄
Molecular Weight: 201,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203343
CAS-Nummer: 1029716-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR