4-(2-Fluoroethoxy)phenylboronic acid, pinacol ester 97%
Lieferant: Apollo Scientific
Synonyme:
2-(4-(2-Fluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PC412417-250MG
APOSPC412417-250MGEA
112
EUR
APOSPC412417-250MG
4-(2-Fluoroethoxy)phenylboronic acid, pinacol ester 97%
4-(2-Fluoroethoxy)phenylboronic acid, pinacol ester
Formel:
C₁₄H₂₀BFO₃ Molecular Weight: 266.116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08706126 CAS-Nummer: 864754-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...