[2-Fluoro-4-(hydroxymethyl)phenyl]boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-4-(hydroxymethyl)phenyl)boronic acid, 2-Fluoro-4-(hydroxymethyl)benzeneboronic acid

PC412422-1G PC412422-5G
APOSPC412422-1GEA 125 EUR
APOSPC412422-1G APOSPC412422-5G
[2-Fluoro-4-(hydroxymethyl)phenyl]boronic acid
[2-Fluoro-4-(hydroxymethyl)phenyl]boronic acid
Formel: C₇H₈BFO₃
Molecular Weight: 169.946 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21603758
CAS-Nummer: 1331945-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR