5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 5-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester

PC412439-1G PC412439-250MG PC412439-5G
APOSPC412439-1GEA 122 EUR
APOSPC412439-1G APOSPC412439-250MG APOSPC412439-5G
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 97%
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
Formel: C₁₄H₁₈BFO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16996304
CAS-Nummer: 1400976-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR