2-Fluoro-6-methylphenylboronic acid

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-6-methylphenyl)boronic acid

PC412449-1G
APOSPC412449-1GEA 104 EUR
APOSPC412449-1G
2-Fluoro-6-methylphenylboronic acid
2-Fluoro-6-methylphenylboronic acid
Formel: C₇H₈BFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13194360
CAS-Nummer: 887471-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR