6-Fluoropyridine-2-boronic acid 95+%
Lieferant: Apollo Scientific
Synonyme:
(6-Fluoropyridin-2-yl)boronic acid
PC412475-250MG
APOSPC412475-250MGEA
116
EUR
APOSPC412475-250MG
6-Fluoropyridine-2-boronic acid 95+%
6-Fluoropyridine-2-boronic acid
Formel:
C₅H₅BFNO₂ Molecular Weight: 140,908 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08705713 CAS-Nummer: 916176-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...