2-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-(2-Fluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

PC412493-1G
APOSPC412493-1GEA 72.5 EUR
APOSPC412493-1G
2-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester 95+%
2-Fluor-4-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BF₄O₂
Molecular Weight: 290,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458187
CAS-Nummer: 1073353-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR