2-Hydroxymethyl-5-(trifluoromethyl)phenylboronic acid

Lieferant: Apollo Scientific

PC412496-1G
APOSPC412496-1GEA 1320 EUR
APOSPC412496-1G
2-Hydroxymethyl-5-(trifluoromethyl)phenylboronic acid
2-Hydroxymethyl-5-(trifluoromethyl)phenylboronic acid
Formel: C₈H₈BF₃O₃
Molecular Weight: 219,954 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20526380
CAS-Nummer: 174671-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR