2-Hydroxy-4-(trifluormethyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (2-Hydroxy-4-(trifluoromethyl)phenyl)boronic acid

PC412500-1G PC412500-500MG PC412500-5G PC412500-25G
APOSPC412500-1GEA 87 EUR
APOSPC412500-1G APOSPC412500-500MG APOSPC412500-5G APOSPC412500-25G
2-Hydroxy-4-(trifluormethyl)phenylboronsäure 97%
2-Hydroxy-4-(trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205.9270 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094996
CAS-Nummer: 1072951-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR