3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific

PC412508-250MG
APOSPC412508-250MGEA 149 EUR
APOSPC412508-250MG
3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester 95%
3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester
Formel: C₁₅H₁₈BF₃O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD12026096
CAS-Nummer: 1150561-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR