2-(2,2,2-Trifluoroethoxy)pyrimidine-5-boronic acid
Lieferant: Apollo Scientific
PC412544-250MG
APOSPC412544-250MGEA
118
EUR
APOSPC412544-250MG
2-(2,2,2-Trifluoroethoxy)pyrimidine-5-boronic acid
2-(2,2,2-Trifluoroethoxy)pyrimidine-5-boronic acid
Formel:
C₆H₆BF₃N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375137 CAS-Nummer: 1401163-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...