[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (2-Chloro-6-(trifluoromethyl)pyridin-4-yl)boronic acid, 2-Chloro-6-(trifluoromethyl)pyridine-4-boronic acid

PC412554-1G PC412554-250MG
APOSPC412554-1GEA 179 EUR
APOSPC412554-1G APOSPC412554-250MG
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid 95%
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid
Formel: C₆H₄BClF₃NO₂
Molecular Weight: 225.361 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188845
CAS-Nummer: 1446486-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR