8-(Trifluormethyl)chinolin-6-boronsäure

Lieferant: Apollo Scientific

PC412567-1G PC412567-250MG
APOSPC412567-1GEA 406 EUR
APOSPC412567-1G APOSPC412567-250MG
8-(Trifluormethyl)chinolin-6-boronsäure
8-(Trifluormethyl)chinolin-6-boronsäure
Formel: C₁₀H₇BF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11504852
CAS-Nummer: 1072951-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR