5-Chloro-2-(difluoromethoxy)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC4127-250MG
APOSPC4127-250MGEA
453
EUR
APOSPC4127-250MG
5-Chloro-2-(difluoromethoxy)benzaldehyde 95%
5-Chloro-2-(difluoromethoxy)benzaldehyde
Formel:
C₈H₅ClF₂O₂ Molecular Weight: 206,574 g/mol Siedepunkt: 82…83 °C (0,03 mm Hg) Schmelzpunkt: 14…16 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412195 CAS-Nummer: 145742-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...