1-[Chloro(difluoro)methoxy]-4-nitrobenzene 97%

Lieferant: Apollo Scientific
Achtung

PC4134-1G PC4134-500MG PC4134-5G
APOSPC4134-1GEA 58 EUR
APOSPC4134-1G APOSPC4134-500MG APOSPC4134-5G
1-[Chloro(difluoro)methoxy]-4-nitrobenzene 97%
1-[Chloro(difluoro)methoxy]-4-nitrobenzene
Formel: C₇H₄ClF₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01820808
CAS-Nummer: 40750-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR