6-Fluoro-2-methyl-1,2,3,4-tetrahydroquinoline 98%

Lieferant: Apollo Scientific

PC4146-1G
APOSPC4146-1GEA 53.8 EUR
APOSPC4146-1G
6-Fluoro-2-methyl-1,2,3,4-tetrahydroquinoline 98%
6-Fluoro-2-methyl-1,2,3,4-tetrahydroquinoline
Formel: C₁₀H₁₂FN
Molecular Weight: 165,2100 g/mol
Schmelzpunkt: 32…34 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040976
CAS-Nummer: 42835-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR