3-Fluor-DL-phenylalanin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH
PC4148-250MG
PC4148-5G
PC4148-25G
APOSPC4148-250MGEA
21.9
EUR
APOSPC4148-250MG
APOSPC4148-5G
APOSPC4148-25G
3-Fluor-DL-phenylalanin 98%
3-Fluor-DL-phenylalanin
Formel:
C₉H₁₀FNO₂ Schmelzpunkt: 240…250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004273 CAS-Nummer: 456-88-2 EINECS: 220-104-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...