4-Fluorbenzylidenaceton 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Fluorophenyl)but-1-en-3-one
PC4150-1G
APOSPC4150-1GEA
219
EUR
APOSPC4150-1G
4-Fluorbenzylidenaceton 97%
4-Fluorbenzylidenaceton
Formel:
C₁₀H₉FO Siedepunkt: 110…112 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205144 CAS-Nummer: 1611-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...