5-Chloro-2,3-difluoro-4-methylbenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC4151-1G
APOSPC4151-1GEA 0 EUR
APOSPC4151-1G
5-Chloro-2,3-difluoro-4-methylbenzoic acid 97%
5-Chloro-2,3-difluoro-4-methylbenzoic acid
Formel: C₈H₅ClF₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03701034
CAS-Nummer: 773869-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR