4-(Chlordifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC4153-1G
APOSPC4153-1GEA 643 EUR
APOSPC4153-1G
4-(Chlordifluormethyl)benzonitril 98%
4-(Chlordifluormethyl)benzonitril
Formel: C₈H₄ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD04972712
CAS-Nummer: 51012-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR