4-Fluorophenyl-2-thienyl ketone 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC4161-10G
APOSPC4161-10GEA
109
EUR
APOSPC4161-10G
4-Fluorophenyl-2-thienyl ketone 99%
4-Fluorophenyl-2-thienyl ketone
Formel:
C₁₁H₇FOS Molecular Weight: 206,23 g/mol Schmelzpunkt: 97…99 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00044554 CAS-Nummer: 579-49-7 EINECS: 209-442-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...