5-(4-Fluorphenyl)oxazol 97%
Lieferant: Apollo Scientific
PC4174C-1G
APOSPC4174C-1GEA
55.1
EUR
APOSPC4174C-1G
5-(4-Fluorphenyl)oxazol 97%
5-(4-Fluorophenyl)oxazole
Formel:
C₉H₆FNO Molecular Weight: 163,1485 g/mol |
MDL:
MFCD00085087 CAS-Nummer: 128101-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...