2-(4-Fluorphenyl)-2-propanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
alpha,alpha-Dimethyl-4-fluorobenzyl alcohol, 2-(4-Fluorophenyl)propan-2-ol
PC4174E-1G
APOSPC4174E-1GEA
21.9
EUR
APOSPC4174E-1G
2-(4-Fluorphenyl)-2-propanol 97%
2-(4-Fluorophenyl)-2-propanol
Formel:
C₉H₁₁FO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059919 CAS-Nummer: 402-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...