3-(4-Fluorphenyl)propionsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC4174G-1G
PC4174G-5G
PC4174G-10G
APOSPC4174G-1GEA
21.9
EUR
APOSPC4174G-1G
APOSPC4174G-5G
APOSPC4174G-10G
3-(4-Fluorphenyl)propionsäure 97%
3-(4-Fluorphenyl)propionsäure
Formel:
C₉H₉FO₂ Molecular Weight: 168.17 g/mol Schmelzpunkt: 86…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060327 CAS-Nummer: 459-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...