1-(4-Fluorphenyl)pyrrol 97%

Lieferant: Apollo Scientific

PC4174M-1G PC4174M-5G
APOSPC4174M-1GEA 61.3 EUR
APOSPC4174M-1G APOSPC4174M-5G
1-(4-Fluorphenyl)pyrrol 97%
1-(4-Fluorophenyl)pyrrole
Formel: C₁₀H₈FN
Schmelzpunkt: 52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047066
CAS-Nummer: 81329-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR