1-Fluor-4-(trifluormethylthio)benzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluorophenyl trifluoromethyl sulphide
PC4176-1G
APOSPC4176-1GEA
132
EUR
APOSPC4176-1G
1-Fluor-4-(trifluormethylthio)benzol 98%
1-Fluoro-4-(trifluoromethylthio)benzene
Formel:
C₇H₄F₄S Molecular Weight: 196.17 g/mol Siedepunkt: 136…138 °C Flammpunkt: 37 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00040978 CAS-Nummer: 940-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...