4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC4182-1G PC4182-5G
APOSPC4182-1GEA 0 EUR
APOSPC4182-1G APOSPC4182-5G
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
Formel: C₁₀H₆F₆O₃
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236043
CAS-Nummer: 16261-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR