2-Hydroxy-4-(trifluoromethyl)pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Trifluoromethyl)-2-pyrimidinol

PC4184-5G
APOSPC4184-5GEA 21.9 EUR
APOSPC4184-5G
2-Hydroxy-4-(trifluoromethyl)pyrimidine 97%
2-Hydroxy-4-(trifluoromethyl)pyrimidine
Formel: C₅H₃F₃N₂O
Molecular Weight: 164,09 g/mol
Schmelzpunkt: 223…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192529
CAS-Nummer: 104048-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR