5-Chloro-2,4-difluoro-6-methylpyrimidine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2,4-difluoro-6-methyl-1,3-diazine

PC4191-1G
APOSPC4191-1GEA 234 EUR
APOSPC4191-1G
5-Chloro-2,4-difluoro-6-methylpyrimidine 98%
5-Chloro-2,4-difluoro-6-methylpyrimidine
Formel: C₅H₃ClF₂N₂
Molecular Weight: 164,541 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036058
CAS-Nummer: 72630-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR