3-[(Chlorodifluoromethyl)thio]pentane-2,4-dione 98%

Lieferant: Apollo Scientific

Synonyme: 3-[(Chlorodifluoromethyl)sulphanyl]pentane-2,4-dione

PC4192-1G
APOSPC4192-1GEA 655 EUR
APOSPC4192-1G
3-[(Chlorodifluoromethyl)thio]pentane-2,4-dione 98%
3-[(Chlorodifluoromethyl)thio]pentane-2,4-dione
Formel: C₆H₇ClF₂O₂S
Molecular Weight: 216,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059533
CAS-Nummer: 42092-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR