Methyl-3,3-difluorocyclobutanecarboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3,3-difluorocyclobutanecarboxylate, Methyl 3,3-difluorocyclobutane-1-carboxylate

PC420000-25G
APOSPC420000-25GEA 215 EUR
APOSPC420000-25G
Methyl-3,3-difluorocyclobutanecarboxylate 98%
Methyl-3,3-difluorocyclobutanecarboxylate
Formel: C₆H₈F₂O₂
Molecular Weight: 150.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015922
CAS-Nummer: 1234616-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR